missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 8 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8458-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055217.2 | |
Cytochrome b-c1 complex subunit 8 Protein | |
Research Use Only | |
QP-C/UQCRQ | |
Recombinant Protein | |
E. coli | |
GREFGNLTRM RHVISYSLSP FEQRAYPHVF TKGIPNVLRR IRESFFRVVP QFVVFYLIYT WGTEEFERSK RKNPAAY | |
Greater than 90% as determined by SDS-PAGE. |
27089 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |