missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cysteine dioxygenase type 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Cysteine dioxygenase type 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7589-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089590

  • 3450.00 DKK / 100µg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001310494.1
Cysteine dioxygenase type 1 Protein
Research Use Only
CDO1
Recombinant Protein
E. coli
MEQTEVLKPR TLADLIRILH QLFAGDEVNV EEVQAIMEAY ESDPTEWAMY AKFDQYRYTR NLVDQGNGKF NLMILCWGEG HGSSIHDHTN SHCFLKMLQG NLKETLFAWP DKKSNEMVKK SERVLRENQC AYINDSIGLH RVENISHTEP AVSLHLYSPP FDTCHAFDQR TGHKNKVTMT FHSKFGIRTP NATSGSLENN
Greater than 80% as determined by SDS-PAGE.
1036
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers