missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cystatin-9 Protein
A DNA sequence encoding the Homo sapiens (Human) Cystatin-9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7699-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008693.2 | |
Cystatin-9 Protein | |
Research Use Only | |
CST9 | |
Recombinant Protein | |
E. coli | |
WCSEEEMGGN NKIVQDPMFL ATVEFALNTF NVQSKEEHAY RLLRVLSSWR EDSMDRKWRG KMVFSMNLQL RQTVCRKFED DIDNCPFQES LELNNVRQGI SFPQVHSCGC CMGCGVGTGA ADKAIPRDKG K | |
Greater than 90% as determined by SDS-PAGE. |
128822 | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |