missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Crossover junction endonuclease EME1 Protein

A DNA sequence encoding the Homo sapiens (Human) Crossover junction endonuclease EME1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8003-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092091

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001159603.1
Crossover junction endonuclease EME1 Protein
Research Use Only
EME1
Recombinant Protein
E. coli
MALKKSSPSL DSGDSDSEEL PTFAFLKKEP SSTKRRQPER EEKIVVVDIS DCEASCPPAP ELFSPPVPEI AETVTQTQPV RLLSSESEDE EEFIPLAQRL TCKFLTHKQL SPEDSSSPVK SVLDHQNNEG ASCDWKKPFP KIPEVPLHDT PERSAADNKD LILDPCCQLP AYLSTCPGQS SSLAVTKTNS DILPPQKKTK PSQKVQGRGS HGCRQQRQAR QKESTLRRQE RKNAALVTRM KAQRPEECLK HIIVVLDPVL LQMEGGGQLL GALQTMECRC VIEAQAVPCS VTWRRRAGPS EDREDWVEEP TVLVLLRAEA FVSMIDNGKQ GSLDSTMKGK ETLQGFVTDI TAKTAGKALS LVIVDQEKCF SLELLFFDFL PCTSAQNPPR RGKQGANKQT KKQQQRQPEA SIGSMVSRVD AEEALVDLQL HTEAQAQIVQ SWKELADFTC AFTKAVAEAP FKKLRDETTF SFCLESDWAG GVKVDLAGRG LALVWRRQIQ QLNRVSLEMA SAVVNAYPSP QLLVQAYQQC FSDKERQNLL ADIQVRRGEG VTSTSRRIGP ELSRRIYLQM TTLQPHLSLD SAD
Greater than 90% as determined by SDS-PAGE.
146956
200 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers