missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Crossover junction endonuclease EME1 Protein
A DNA sequence encoding the Homo sapiens (Human) Crossover junction endonuclease EME1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8003-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001159603.1 | |
Crossover junction endonuclease EME1 Protein | |
Research Use Only | |
EME1 | |
Recombinant Protein | |
E. coli | |
MALKKSSPSL DSGDSDSEEL PTFAFLKKEP SSTKRRQPER EEKIVVVDIS DCEASCPPAP ELFSPPVPEI AETVTQTQPV RLLSSESEDE EEFIPLAQRL TCKFLTHKQL SPEDSSSPVK SVLDHQNNEG ASCDWKKPFP KIPEVPLHDT PERSAADNKD LILDPCCQLP AYLSTCPGQS SSLAVTKTNS DILPPQKKTK PSQKVQGRGS HGCRQQRQAR QKESTLRRQE RKNAALVTRM KAQRPEECLK HIIVVLDPVL LQMEGGGQLL GALQTMECRC VIEAQAVPCS VTWRRRAGPS EDREDWVEEP TVLVLLRAEA FVSMIDNGKQ GSLDSTMKGK ETLQGFVTDI TAKTAGKALS LVIVDQEKCF SLELLFFDFL PCTSAQNPPR RGKQGANKQT KKQQQRQPEA SIGSMVSRVD AEEALVDLQL HTEAQAQIVQ SWKELADFTC AFTKAVAEAP FKKLRDETTF SFCLESDWAG GVKVDLAGRG LALVWRRQIQ QLNRVSLEMA SAVVNAYPSP QLLVQAYQQC FSDKERQNLL ADIQVRRGEG VTSTSRRIGP ELSRRIYLQM TTLQPHLSLD SAD | |
Greater than 90% as determined by SDS-PAGE. |
146956 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |