missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COP9 signalosome complex subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) COP9 signalosome complex subunit 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7761-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001137359.1 | |
COP9 signalosome complex subunit 2 Protein | |
Research Use Only | |
COPS2 | |
Recombinant Protein | |
E. coli | |
MSDMEDDFMC DDEEDYDLEY SEDSNSEPNV DLENQYYNSK ALKEDDPKAA LSSFQKVLEL EGEKGEWGFK ALKQMIKINF KLTNFPEMMN RYKQLLTYIR SAVTRNYSEK SINSILDYIS TSKQMDLLQE FYETTLEALK DAKNDRLWFK TNTKLGKLYL EREEYGKLQK ILRQLHQSCQ TDDGEDDLKK GTQLLEIYAL EIQMYTAQKN NKKLKALYEQ SLHIKSAIPH PLIMGVIREC GGKMHLREGE FEKAHTDFFE AFKNYDESGS PRRTTCLKYL VLANMLMKSG INPFDSQEAK PYKNDPEILA MTNLVSAYQN NDITEFEKIL KTNHSNIMDD PFIREHIEEL LRNIRTQVLI KLIKPYTRIH IPFISKELNI DVADVESLLV QCILDNTIHG RIDQVNQLLE LDHQKRGGAR YTALDKWTNQ LNSLNQAVVS KLA | |
Greater than 90% as determined by SDS-PAGE. |
9318 | |
10 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human COP9 signalosome complex subunit 2 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction