missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COP9 signalosome complex subunit 2 Protein

A DNA sequence encoding the Homo sapiens (Human) COP9 signalosome complex subunit 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7761-ec-500ug

 View more versions of this product

Product Code. 16010651

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001137359.1
COP9 signalosome complex subunit 2 Protein
Research Use Only
COPS2
Recombinant Protein
E. coli
MSDMEDDFMC DDEEDYDLEY SEDSNSEPNV DLENQYYNSK ALKEDDPKAA LSSFQKVLEL EGEKGEWGFK ALKQMIKINF KLTNFPEMMN RYKQLLTYIR SAVTRNYSEK SINSILDYIS TSKQMDLLQE FYETTLEALK DAKNDRLWFK TNTKLGKLYL EREEYGKLQK ILRQLHQSCQ TDDGEDDLKK GTQLLEIYAL EIQMYTAQKN NKKLKALYEQ SLHIKSAIPH PLIMGVIREC GGKMHLREGE FEKAHTDFFE AFKNYDESGS PRRTTCLKYL VLANMLMKSG INPFDSQEAK PYKNDPEILA MTNLVSAYQN NDITEFEKIL KTNHSNIMDD PFIREHIEEL LRNIRTQVLI KLIKPYTRIH IPFISKELNI DVADVESLLV QCILDNTIHG RIDQVNQLLE LDHQKRGGAR YTALDKWTNQ LNSLNQAVVS KLA
Greater than 90% as determined by SDS-PAGE.
9318
500 ÎĽg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human COP9 signalosome complex subunit 2 Protein > 500ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.