Få mere at vide
R&D Systems™ Recombinant Human Coagulation Factor III/Tissue Factor
Mængde:
10 μg
Beskrivelse
The Recombinant Human Coagulation Factor III/Tissue Factor is derived from NS0. The Recombinant Human Coagulation Factor III/Tissue Factor has been validated for the following applications: Enzyme Activity.
Tekniske data
Tekniske data
| Formulering | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gen-id (Entrez) | 2152 |
| Molekylvægt (g/mol) | 26 kDa |
| Navn | Coagulation Factor III/Tissue Factor |
| Mængde | 10 μg |
| Kilde | Mouse myeloma cell line,NS0-derived human Coagulation Factor III/Tissue Factor protein Gly34-Glu251,with a C-terminal 6-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |