Få mere at vide
R&D Systems™ Recombinant Human Coagulation Factor II/Thrombin Protein
Artikelnummer. 18107190
Mængde:
10 μg
Artikelnummer. 18107190
Brand: R&D Systems™ 1473SE010
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
The Recombinant Human Coagulation Factor II/Thrombin Protein is derived from NS0. The Recombinant Human Coagulation Factor II/Thrombin Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY
Tekniske data
Formulering Lyophilized from a 0.2μm filtered solution in HEPES and NaCl.
Gen-id (Entrez) 2147
Molekylvægt (g/mol) 4 kDa (light chain A), 31 kDa (heavy chain B), 14 kDa (C-terminal fragment)
Navn Coagulation Factor II/Thrombin
Mængde 10 μg
Kilde Mouse myeloma cell line,NS0-derived human Coagulation Factor II/Thrombin protein Met1-Glu622 with a C-terminal 10-His tag The proenzyme was purified,activated and further purified.
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Active form
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.