missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Coactosin-like Protein

A DNA sequence encoding the Homo sapiens (Human) Coactosin-like protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7594-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089620

  • 3455.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_066972.1
Coactosin-like Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
23406
Greater than 90% as determined by SDS-PAGE.
Research Use Only
COTL1
Recombinant Protein
E. coli
ATKIDKEACR AAYNLVRDDG SAVIWVTFKY DGSTIVPGEQ GAEYQHFIQQ CTDDVRLFAF VRFTTGDAMS KRSKFALITW IGENVSGLQR AKTGTDKTLV KEVVQNFAKE FVISDRKELE EDFIKSELKK AGGANYDAQT E
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers