missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CINP Protein
A DNA sequence encoding the Homo sapiens (Human) CINP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8044-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001306975.1 | |
CINP Protein | |
Research Use Only | |
CINP | |
Recombinant Protein | |
E. coli | |
MEAKTLGTVT PRKPVLSVSA RKIKDNAADW HNLILKWETL NDAGFTTANN IANLKISLLN KDKIELDSSS PASKENEEKV CLEYNEELEK LCEELQATLD GLTKIQVKME KLSSTTKGIC ELENYHYGEE SKRPPLFHTW PTTHFYEVSH KLLEMYRKEL LLKRTVAKEL AHTGDPDLTL SYLSMWLHQP YVESDSRLHL ESMLLETGHR AL | |
Greater than 90% as determined by SDS-PAGE. |
51550 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |