Learn More
enQuireBio™ Recombinant Human CHD1L Protein
A DNA sequence encoding the Homo sapiens (Human) CHD1L, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7802-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16070881
  • 5015.00 DKK / 100µg
Estimated Shipment: 05-07-2024
to see stock.
Specifications
NP_001243267.1
CHD1L Protein
Research Use Only
CHD1L
Recombinant Protein
E. coli
SAELDYQDPD ATSLKYVSGD VTHPQAGAED ALIVHCVDDS GHWGRGGLFT ALEKRSAEPR KIYELAGKMK DLSLGGVLLF PVDDKESRNK GQDLLALIVA QHRDRSNVLS GIKMAALEEG LKKIFLAAKK KKASVHLPRI GHATKGFNWY GTERLIRKHL AARGIPTYIY YFPRSKSAVL HAQSSSSSSR QLVP
Greater than 90% as determined by SDS-PAGE.
9557
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol