missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHD1L Protein
A DNA sequence encoding the Homo sapiens (Human) CHD1L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7802-ec-50ug
Additional Details : Weight : 0.01000kg
Specifica
NP_001243267.1 | |
CHD1L Protein | |
Research Use Only | |
CHD1L | |
Recombinant Protein | |
E. coli | |
SAELDYQDPD ATSLKYVSGD VTHPQAGAED ALIVHCVDDS GHWGRGGLFT ALEKRSAEPR KIYELAGKMK DLSLGGVLLF PVDDKESRNK GQDLLALIVA QHRDRSNVLS GIKMAALEEG LKKIFLAAKK KKASVHLPRI GHATKGFNWY GTERLIRKHL AARGIPTYIY YFPRSKSAVL HAQSSSSSSR QLVP | |
Greater than 90% as determined by SDS-PAGE. |
9557 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |