missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human CD45 / PTPRC Protein

Artikelnummer. p-7155704
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16098171

missing translation for 'mfr': enQuireBio™ QP9356ye10ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) CD45 / PTPRC, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_002829.3
Gen-id (Entrez) 5788
Navn CD45 / PTPRC Protein
Mængde 10 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol PTPRC
Protein længde Extracellular Domain
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter Yeast
Protein tag His
Sekvens QSPTPSPTGL TTAKMPSVPL SSDPLPTHTT AFSPASTFER ENDFSETTTS LSPDNTSTQV SPDSLDNASA FNTTGVSSVQ TPHLPTHADS QTPSAGTDTQ TFSGSAANAK LNPTPGSNAI SDVPGERSTA STFPTDPVSP LTTTLSLAHH SSAALPARTS NTTITANTSD AYLNASETTT LSPSGSAVIS TTTIATTPSK PTCDEKYANI TVDYLYNKET KLFTAKLNVN ENVECGNNTC TNNEVHNLTE CKNASVSISH NSCTAPDKTL ILDVPPGVEK FQLHDCTQVE KADTTICLKW KNIETFTCDT QNITYRFQCG NMIFDNKEIK LENLEPEHEY KCDSEILYNN HKFTNASKII KTDFGSPGEP QIIFCRSEAA HQGVITWNPP QRSFHNFTLC YIKETEKDCL NLDKNLIKYD LQNLKPYTKY VLSLHAYIIA KVQRNGSAAM CHFTTKSAPP SQVWNMTVSM TSDNSMHVKC RPPRDRNGPH ERYHLEVEAG NTLVRNESHK NCDFRVKDLQ YSTDYTFKAY FHNGDYPGEP FILHHSTSYN SK
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.