Få mere at vide
R&D Systems™ Recombinant Human Cathepsin B Protein
Beskrivelse
The Recombinant Human Cathepsin B Protein is derived from NS0. The Recombinant Human Cathepsin B Protein has been validated for the following applications: Enzyme Activity.
Tekniske data
Tekniske data
| Formulering | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
| Gen-id (Entrez) | 1508 |
| Molekylvægt (g/mol) | 37 kDa (Pro) & 29 kDa (Mature) |
| Navn | Cathepsin B |
| Mængde | 10 μg |
| Kilde | Mouse myeloma cell line,NS0-derived human Cathepsin B protein Arg18-Ile339 (pro) & Phe74-Ile339 (mature),both with a C-terminal 10-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Strukturel form | Pro and Mature forms |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |