Få mere at vide
Novus Biologicals™ Recombinant Human Cardiotrophin-1/CT-1 Protein
Artikelnummer. 18031029 Shop alle Bio Techne produkter
Skift visning
Mængde:
50 μg
Artikelnummer. Mængde
18031029 50 μg
1 options
Artikelnummer. 18031029 Leverandør Novus Biologicals™ Leverandørnr. NBP19974550UG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results.
Recombinant protein containing 200 amino acids for Human Cardiotrophin-1/CT-1 The Recombinant Human Cardiotrophin-1/CT-1 Protein is derived from E. coli. The Recombinant Human Cardiotrophin-1/CT-1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Tekniske data
Koncentration Lyoph
Til brug med (applikation) SDS-PAGE
Formulering Sterile filtered and lyophilized from 1X PBS.
Gen-id (Entrez) 1489
Molekylvægt (g/mol) 22.5 kDa
Mængde 50 μg
Kilde E. coli
Opbevaringskrav Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1
Gen symbol CTF1
Rekonstitution Reconstitute in 20 mM Tris, pH 8.0 to a concentration of <1.0 mg/mL. The solution can be diluted into other aqueous buffers.
Buffer Sterile filtered and lyophilized from 1X PBS.
Endotoksin niveau <0.1 ng/μg
Renhed eller kvalitetsklasse >98%, by SDS-PAGE