Få mere at vide
R&D Systems™ Recombinant Human Carbonic Anhydrase III Protein
Mængde:
50 μg
Beskrivelse
The Recombinant Human Carbonic Anhydrase III Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase III Protein has been validated for the following applications: Enzyme Activity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
| Gen-id (Entrez) | 761 |
| Molekylvægt (g/mol) | 31 kDa |
| Navn | Carbonic Anhydrase III/CA3 |
| Mængde | 50 μg |
| Kilde | E. coli-derived human Carbonic Anhydrase III/CA3 protein Ala2-Lys260 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |