missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-2/BMP-6 Heterodimer Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
Description
The Recombinant Human BMP-2/BMP-6 Heterodimer Protein is derived from E. coli. The Recombinant Human BMP-2/BMP-6 Heterodimer Protein has been validated for the following applications: Bioactivity.

Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in HCl. |
Gene ID (Entrez) | 650 |
Molecular Weight (g/mol) | 12.8 kDa (BMP-2) & 15 kDa (BMP-6) |
Name | BMP-2/BMP-6 Heterodimer |
Quantity | 10 μg |
Source | E. coli-derived human BMP-2/BMP-6 Heterodimer protein, (N-terminus) Human BMP-2 (Ala284-Arg396),with an N-terminal Met Accession &Num; P12643 Human BMP-6 (Gln382-His513),with an N-terminal Met Accession &Num; P22004 (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked heterodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human BMP-2/BMP-6 Heterodimer Protein >
Spot an opportunity for improvement?Share a Content Correction