missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-catenin-interacting protein 1

A DNA sequence encoding the Homo sapiens (Human) Beta-catenin-interacting protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8120-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052771

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001012329.1
Beta-catenin-interacting protein 1
Research Use Only
CTNNBIP1
Recombinant Protein
E. coli
MNREGAPGKS PEEMYIQQKV RVLLMLRKMG SNLTASEEEF LRTYAGVVNS QLSQLPPHSI DQGAEDVVMA FSRSETEDRR Q
Greater than 80% as determined by SDS-PAGE.
56998
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers