missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-1 adrenergic receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-1 adrenergic receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8958-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_000675.1 | |
Beta-1 adrenergic receptor Protein | |
Research Use Only | |
ADRB1 | |
Recombinant Protein | |
Yeast | |
CRSPDFRKAF QRLLCCARRA ARRRHATHGD RPRASGCLAR PGPPPSPGAA SDDDDDDVVG ATPPARLLEP WAGCNGGAAA DSDSSLDEPC RPGFASESKV | |
Greater than 90% as determined by SDS-PAGE. |
153 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Beta-1 adrenergic receptor Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction