missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP6V1F Protein

A DNA sequence encoding the Homo sapiens (Human) ATP6V1F, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP909-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047621

  • 10455.00 DKK / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001185838.1
ATP6V1F Protein
Research Use Only
ATP6V1F
Recombinant Protein
E. coli
MAGRGKLIAV IGDEDTVTGF LLGGIGELNK NRHPNFLVVE KDTTINEIED TFRQFLNRDD IGIILINQYI AEMVRHALDA HQQSIPAVLE IPSKEHPYDA AKDSILRRAR GMFTAEDLR
Greater than 90% as determined by SDS-PAGE.
9296
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers