missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Astrocytic phosphoprotein PEA-15 Protein
A DNA sequence encoding the Homo sapiens (Human) Astrocytic phosphoprotein PEA-15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7581-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001284505.1 | |
Astrocytic phosphoprotein PEA-15 Protein | |
Research Use Only | |
PEA15 | |
Recombinant Protein | |
E. coli | |
MAEYGTLLQD LTNNITLEDL EQLKSACKED IPSEKSEEIT TGSAWFSFLE SHNKLDKDNL SYIEHIFEIS RRPDLLTMVV DYRTRVLKIS EEDELDTKLT RIPSAKKYKD IIRQPSEEEI IKLAPPPKKA | |
Greater than 80% as determined by SDS-PAGE. |
8682 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |