missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Tekniske data
Tekniske data
| Til brug med (applikation) | Western Blot, ELISA, Immunoprecipitation, Protein Array, Immunoaffinity Purification |
| Formulering | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-id (Entrez) | 55738 |
| Molekylvægt (g/mol) | MolecularWeight-theroretical: 71.1 kDa |
| Mængde | 10 μg |
| Opbevaringskrav | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924 |
| Gen symbol | ARFGAP1 |
| Inhibitor egenskaber | Use in Functional Assay reported in scientific literature (PMID: 24282027). |
| Renhed eller kvalitetsklasse | >80% by SDS-PAGE and Coomassie blue staining |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?