Beskrivelse
Recombinant protein containing 300 amino acids for Human ApoE4
The Recombinant Human ApoE4 Protein is derived from E. coli. The Recombinant Human ApoE4 Protein has been validated for the following applications: Western Blot, SDS-Page.
The Recombinant Human ApoE4 Protein is derived from E. coli. The Recombinant Human ApoE4 Protein has been validated for the following applications: Western Blot, SDS-Page.
Tekniske data
Tekniske data
| Koncentration | Lyoph |
| Til brug med (applikation) | Western Blot, SDS-PAGE |
| Gen-id (Entrez) | 348 |
| Molekylvægt (g/mol) | 34.4 kDa |
| Mængde | 1000 μg |
| Kilde | E. coli |
| Opbevaringskrav | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <0.1 ng/ug |
| Gen symbol | APOE |
| Vis mere |