missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA6 / Annexin A6 / Annexin6 Protein
A DNA sequence encoding the Homo sapiens (Human) ANXA6 / Annexin A6 / Annexin6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8334-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001146.2 | |
ANXA6 / Annexin A6 / Annexin6 Protein | |
Research Use Only | |
ANXA6 | |
Recombinant Protein | |
E. coli | |
AKPAQGAKYR GSIHDFPGFD PNQDAEALYT AMKGFGSDKE AILDIITSRS NRQRQEVCQS YKSLYGKDLI ADLKYELTGK FERLIVGLMR PPAYCDAKEI KDAISGIGTD EKCLIEILAS RTNEQMHQLV AAYKDAYERD LEADIIGDTS GHFQKMLVVL LQGTREEDDV VSEDLVQQDV QDLYEAGELK WGTDEAQFIY ILGNRSKQHL RLVFDEYLKT TGKPIEASIR GELSGDFEKL MLAV | |
Greater than 90% as determined by SDS-PAGE. |
309 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |