missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANXA6 / Annexin A6 / Annexin6 Protein

A DNA sequence encoding the Homo sapiens (Human) ANXA6 / Annexin A6 / Annexin6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8334-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044071

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001146.2
ANXA6 / Annexin A6 / Annexin6 Protein
Research Use Only
ANXA6
Recombinant Protein
E. coli
AKPAQGAKYR GSIHDFPGFD PNQDAEALYT AMKGFGSDKE AILDIITSRS NRQRQEVCQS YKSLYGKDLI ADLKYELTGK FERLIVGLMR PPAYCDAKEI KDAISGIGTD EKCLIEILAS RTNEQMHQLV AAYKDAYERD LEADIIGDTS GHFQKMLVVL LQGTREEDDV VSEDLVQQDV QDLYEAGELK WGTDEAQFIY ILGNRSKQHL RLVFDEYLKT TGKPIEASIR GELSGDFEKL MLAV
Greater than 90% as determined by SDS-PAGE.
309
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers