missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-related protein 2/3 complex subunit 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8442-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_005722.1 | |
Actin-related protein 2 / 3 complex subunit 2 Protein | |
Research Use Only | |
ARPC2 | |
Recombinant Protein | |
E. coli | |
MILLEVNNRI IEETLALKFE NAAAGNKPEA VEVTFADFDG VLYHISNPNG DKTKVMVSIS LKFYKELQAH GADELLKRVY GSFLVNPESG YNVSLLYDLE NLPASKDSIV HQAGMLKRNC FASVFEKYFQ FQEEGKEGEN RAVIHYRDDE TMYVESKKDR VTVVFSTVFK DDDDVVIGKV FMQEFKEGRR ASHTAPQVLF SHREPPLELK DTDAAVGDNI GYITFVLFPR HTNASARDNT INLIHTFRDY | |
Greater than 90% as determined by SDS-PAGE. |
10109 | |
200 ÎĽg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction