missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Acidic mammalian chitinase Protein
A DNA sequence encoding the Homo sapiens (Human) Acidic mammalian chitinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8133-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001035713.1 | |
Acidic mammalian chitinase Protein | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
27159 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
CHIA | |
Recombinant Protein | |
E. coli | |
MVSTPENRQT FITSVIKFLR QYEFDGLDFD WEYPGSRGSP PQDKHLFTVL VQEMREAFEQ EAKQINKPRL MVTAAVAAGI SNIQSGYEIP QLSQYLDYIH VMTYDLHGSW EGYTGENSPL YKYPTDTGSN AYLNVDYVMN YWKDNGAPAE KLIVGFPTYG HNFILSNPSN TGIGAPTSGA GPAGPYAKES GIWAYYEICT FLKNGATQGW DAPQEVPYAY QGNVWVGYDN IKSFDIKAQW LKHNKFGGAM VWAIDLDDFT GTFCNQGKFP LISTLKKALG LQSASCTAPA QPIEPITAAP SGSGNGSGSS SSGGSSGGSG FCAVRANGLY PVANNRNAFW HCVNGVTYQQ NCQAGLVFDT SCDCCNWA |