missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACBD5 Protein

A DNA sequence encoding the Homo sapiens (Human) ACBD5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7710-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001035938.1
ACBD5 Protein
Research Use Only
ACBD5
Recombinant Protein
E. coli
MFQFHAGSWE SWCCCCLIPA DRPWDRGQHW QLEMADTRSV HETRFEAAVK VIQSLPKNGS FQPTNEMMLK FYSFYKQATE GPCKLSRPGF WDPIGRYKWD AWSSLGDMTK EEAMIAYVEE MKKIIETMPM TEKVEELLRV IGPFYEIVED KKSGRSSDIT SVRLEKISKC LEDLGNVLTS TPNAKTVNGK AESSDSGAES EEEEAQEEVK GAEQSDNDKK MMKKSADHKN LEVIVTNGYD KDGFVQDIQN DIHASSSLNG RSTEEVKPID ENLGQTGKSA VCIHQDINDD HVEDVTGIQH LTSDSDSEVY CDSMEQFGQE ESLDSFTSNN GPFQYYLGGH SSQPMENSGF REDIQVPPGN GNIGNMQVVA VEGKGEVKHG GEDGRNNSGA PHREKRGGET DEFSNVRRGR GHRMQHLSEG TKGRQVGSGG DGERWGSDRG SRGSLNEQIA LVLMRLQEDM Q
Greater than 90% as determined by SDS-PAGE.
91452
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers