missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human 60S ribosomal protein L3 Protein

Artikelnummer. 16064431
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16064431

Brand: enQuireBio™ QP8406ec100ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L3, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_000958.1
Gen-id (Entrez) 6122
Navn 60S ribosomal protein L3 Protein
Mængde 100 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol RPL3
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter E. coli
Protein tag GST
Sekvens SHRKFSAPRH GSLGFLPRKR SSRHRGKVKS FPKDDPSKPV HLTAFLGYKA GMTHIVREVD RPGSKVNKKE VVEAVTIVET PPMVVVGIVG YVETPRGLRT FKTVFAEHIS DECKRRFYKN WHKSKKKAFT KYCKKWQDED GKKQLEKDFS SMKKYCQVIR VIAHTQMRLL PLRQKKAHLM EIQVNGGTVA EKLDWARERL EQQVPVNQVF GQDEMIDVIG VTKGKGYKGV TSRWHTKKLP RKTHRG
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.