Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L15 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L15, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8408-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16084441
  • 3450.00 DKK / 100µg
Estimated Shipment: 04-07-2024
to see stock.
Specifications
NP_001240308.1
60S ribosomal protein L15 Protein
Research Use Only
RPL15
Recombinant Protein
E. coli
GAYKYIQELW RKKQSDVMRF LLRVRCWQYR QLSALHRAPR PTRPDKARRL GYKAKQGYVI YRIRVRRGGR KRPVPKGATY GKPVHHGVNQ LKFARSLQSV AEERAGRHCG ALRVLNSYWV GEDSTYKFFE VILIDPFHKA IRRNPDTQWI TKPVHKHREM RGLTSAGRKS RGLGKGHKFH HTIGGSRRAA WRRRNTLQLH RYR
Greater than 90% as determined by SDS-PAGE.
6138
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol