missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S9 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8331-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001004.2 | |
40S ribosomal protein S9 Protein | |
Research Use Only | |
RPS9 | |
Recombinant Protein | |
E. coli | |
PVARSWVCRK TYVTPRRPFE KSRLDQELKL IGEYGLRNKR EVWRVKFTLA KIRKAARELL TLDEKDPRRL FEGNALLRRL VRIGVLDEGK MKLDYILGLK IEDFLERRLQ TQVFKLGLAK SIHHARVLIR QRHIRVRKQV VNIPSFIVRL DSQKHIDFSL RSPYGGGRPG RVKRKNAKKG QGG | |
Greater than 90% as determined by SDS-PAGE. |
6203 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |