Få mere at vide
enQuireBio™ Recombinant hepatitis Non-structural polyprotein pORF1 Protein
Artikelnummer. 16007450
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16007450
Brand: enQuireBio™ QP712550ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Non-structural polyprotein pORF1, was expressed in the hosts and tags indicated.
Tekniske data
Navn hepatitis Non-structural polyprotein pORF1 Protein
Mængde 50 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol ORF1
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Hepatitis
Arter E. coli
Protein tag His
Sekvens EVFWNHPIQR VIHNELELYC RARSGRCLEI GAHPRSINDN PNVVHRCFLR PAGRDVQRWY TAPTRGPAAN CRRSALRGLP AADRTYCFDG FSGCNFPAET GIALYSLHDM SPSDVAEAMF RHGMTRLYAA LHLPPEVLLP PGTYRTASYL LIHDGRRVVV TYEGDTSAGY NHDVSNLRSW I
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.