missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis Capsid Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Capsid protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7715-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
hepatitis Capsid Protein | |
Research Use Only | |
ORF2 | |
Recombinant Protein | |
E. coli | |
PPPGQPSGRR RGRRSGGSGG GFWGDRVDSQ PFAIPYIHPT NPFAPNVTAA AGAGPRVRQP VRPLGSAWRD QAQRPAAASR RRPTTAGAAP LTAVAPAHDT PPVPDVDSRG AILRRQYNLS TSPLTSSVAT GTNLVLYAAP LSPLLPLQDG TNTHIMATEA SNYAQYRVAR ATIRYRPLVP NAVGGYAISI SFWPQTTPTP TSVDMNSITS TDVRILVQPG IASELVIPSE RLHYRNQGWR SVETSGVAEE EATSGLVMLC IHGSPVNSYT NTPYTGALGL LDFALELEFR NLTPGNTNTR VSRYSSTARH RLRRGADGTA ELTTTAATRF MKDLYFTSTN GVGEIGRGIA LTLFNLADTL LGGLPTELIS SAGGQLFYSR PVVSANGEPT VKLYTSVENA QQDKGIAIPN DIDLGESRVV IQDYDNQHEQ DRPTPSPAPS RPFSVLRAND VLWLSLTAAE YDQSTYGSST GPVYVSDSVT LVNVATGAMS -QAVARSLDW TKVTLDGRPL STIQQYSKIF FVLPLRGKLS FWEAGTTRPG YPYNYNTTAS DQLLVENAAG HRVAISTYTT SLGAGPVSIS AVAVLGPHSA LALLEDTLDY PARAHTFDDF CPECRPLGLQ GCAFQSTVAE LQRLKMKVGK TREL | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |