missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C virus genotype 1a (isolate 1) polyProtein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1a (isolate 1) polyprotein, was expressed in the hosts and tags indicated.
2870.00 DKK - 24470.00 DKK
Specifications
Name | Hepatitis C virus genotype 1a (isolate 1) polyProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Partial (see sequence information for more details). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026950
|
enQuireBio™
QP6996-10UG |
10 μg |
2870.00 DKK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056950
|
enQuireBio™
QP6996-20UG |
20 μg |
3695.00 DKK
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046950
|
enQuireBio™
QP6996-200UG |
200 μg |
11525.00 DKK
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066950
|
enQuireBio™
QP6996-500UG |
500 μg |
15400.00 DKK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036950
|
enQuireBio™
QP6996-1MG |
1 mg |
24470.00 DKK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C virus genotype 1a (isolate 1) polyProtein | |
Not determined. | |
Recombinant Protein | |
Mammal | |
YQVRNSTGLY HVTNDCPNSS IVYEAADAIL HTPGCVPCVR EGNASRCWVA MTPTVATRDG KLPATQLRRH IDLLVGSATL CSALYVGDLC GSVFLVGQLF TFSPRRHWTT QGCNCSIYPG HITGHRMAWD MMMN | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Partial (see sequence information for more details). | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |