missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr DNA polymerase catalytic subunit Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) DNA polymerase catalytic subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7276-ec-200ug
This item is not returnable.
View return policy
Specifications
YP_401712.1 | |
DNA polymerase catalytic subunit Protein | |
Research Use Only | |
BALF5 | |
Recombinant Protein | |
E. coli | |
MSGGLFYNPF LRPNKGLLKK PDKEYLRLIP KCFQTPGAAG VVDVRGPQPP LCFYQDSLTV VGGDEDGKGM WWRQRAQEGT ARPEADTHGS PLDFHVYDIL ETVYTHEKCA VIPSDKQGYV VPCGIVIKLL GRRKADGASV CVNVFGQQAY FYASAPQGLD VEFAVLSALK ASTFDRRTPC RVSVEKVTRR SIMGYGNHAG DYHKITLSHP | |
Greater than 90% as determined by SDS-PAGE. |
3783681 | |
200 ÎĽg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Epstein-Barr DNA polymerase catalytic subunit Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction