missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein Product Code.: 16007960

enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein

Product Code. 16007960
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16007960

Brand: enQuireBio™ QP7275ec200ug

Quantity Price
1 9230.00 DKK / 200µg
3 8630.97 DKK / 200µg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) cytomegalovirus (HCMV) Glycoprotein B / gB, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401713.1
Gene ID (Entrez) 3783680
Name cytomegalovirus (HCMV) Glycoprotein B / gB Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol gB
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His-SUMO
Sequence QTPEQPAPPA TTVQPTATRQ QTSFPFRVCE LSSHGDLFRF SSDIQCPSFG TRENHTEGLL MVFKDNIIPY SFKVRSYTKI VTNILIYNGW YADSVTNRHE EKFSVDSYET DQMDTIYQCY NAVKMTKDGL TRVYVDRDGV NITVNLKPTG GLANGVRRYA SQTELYDAPG WLIWTYRTRT TVNCLITDMM AKSNSPFDFF VTTTGQTVEM SPFYDGKNKE TFHERADSFH VRTNYKIV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Epstein-Barr cytomegalovirus (HCMV) Glycoprotein B / gB Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.