missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Translocated intimin receptor Tir Protein
A DNA sequence encoding the Escherichia coli O157:H7 (strain TW14359 / EHEC) Translocated intimin receptor Tir, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7455-ec-200ug
This item is not returnable.
View return policy
Specifications
WP_001301454.1 | |
200 ÎĽg | |
Not determined. | |
Extracellular Domain | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Translocated intimin receptor Tir Protein | |
Research Use Only | |
tir | |
Recombinant Protein | |
E. coli | |
QALALTPEPD SPTTTDPDAA ASATETATRD QLTKEAFQNP DNQKVNIDEL GNAIPSGVLK DDVVANIEEQ AKAAGEEAKQ QAIENNAQAQ KKYDEQQAKR QEELKVSSGA G | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Translocated intimin receptor Tir Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction