missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ruvA Protein
A DNA sequence encoding the Escherichia coli (strain K12) ruvA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7321-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416375.1 | |
ruvA Protein | |
Research Use Only | |
ruvA | |
Recombinant Protein | |
E. coli | |
MIGRLRGIII EKQPPLVLIE VGGVGYEVHM PMTCFYELPE AGQEAIVFTH FVVREDAQLL YGFNNKQERT LFKELIKTNG VGPKLALAIL SGMSAQQFVN AVEREEVGAL VKLPGIGKKT AERLIVEMKD RFKGLHGDLF TPAADLVLTS PASPATDDAE QEAVAALVAL GYKPQEASRM VSKIARPDAS SETLIREALR AAL | |
Greater than 90% as determined by SDS-PAGE. |
946369 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |