missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ruvA Protein

A DNA sequence encoding the Escherichia coli (strain K12) ruvA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7321-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038190

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_416375.1
ruvA Protein
Research Use Only
ruvA
Recombinant Protein
E. coli
MIGRLRGIII EKQPPLVLIE VGGVGYEVHM PMTCFYELPE AGQEAIVFTH FVVREDAQLL YGFNNKQERT LFKELIKTNG VGPKLALAIL SGMSAQQFVN AVEREEVGAL VKLPGIGKKT AERLIVEMKD RFKGLHGDLF TPAADLVLTS PASPATDDAE QEAVAALVAL GYKPQEASRM VSKIARPDAS SETLIREALR AAL
Greater than 90% as determined by SDS-PAGE.
946369
50 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers