Learn More
enQuireBio™ Recombinant E. coli ruvA Protein
A DNA sequence encoding the Escherichia coli (strain K12) ruvA, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7321-ec-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16028190
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
NP_416375.1
ruvA Protein
Research Use Only
ruvA
Recombinant Protein
E. coli
MIGRLRGIII EKQPPLVLIE VGGVGYEVHM PMTCFYELPE AGQEAIVFTH FVVREDAQLL YGFNNKQERT LFKELIKTNG VGPKLALAIL SGMSAQQFVN AVEREEVGAL VKLPGIGKKT AERLIVEMKD RFKGLHGDLF TPAADLVLTS PASPATDDAE QEAVAALVAL GYKPQEASRM VSKIARPDAS SETLIREALR AAL
Greater than 90% as determined by SDS-PAGE.
946369
500 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol