missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein
A DNA sequence encoding the Escherichia coli (strain K12) RNA-splicing ligase RtcB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7212-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_417879.1 | |
RNA-splicing ligase RtcB Protein | |
Research Use Only | |
rtcB | |
Recombinant Protein | |
Yeast | |
MNYELLTTEN APVKMWTKGV PVEADARQQL INTAKMPFIF KHIAVMPDVH LGKGSTIGSV IPTKGAIIPA AVGVDIGCGM NALRTALTAE DLPENLAELR QAIETAVPHG RTTGRCKRDK GAWENPPVNV DAKWAELEAG YQWLTQKYPR FLNTNNYKHL GTLGTGNHFI EICLDESDQV WIMLHSGSRG IGNAIGTYFI DLAQKEMQET LETLPSRDLA YFMEGTEYFD DYLKAVAWAQ LFASLNRDAM MENVVTALQS ITQKTVRQPQ TLAMEEINCH HNYVQKEQHF GEEIYVTRKG AVSARAGQYG IIPGSMGAKS FIVRGLGNEE SFCSCSHGAG RVMSRTKAKK LFSVEDQIRA TAHVECRKDA EVIDEIPMAY KDIDAVMAAQ SDLVEVIYTL RQVVCVKG | |
Greater than 81% as determined by SDS-PAGE. |
947929 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction