missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein Product Code.: 16027740

enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein

Product Code. 16027740
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16027740

Brand: enQuireBio™ QP7212ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) RNA-splicing ligase RtcB, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_417879.1
Gene ID (Entrez) 947929
Name RNA-splicing ligase RtcB Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rtcB
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species Yeast
Protein Tag His
Sequence MNYELLTTEN APVKMWTKGV PVEADARQQL INTAKMPFIF KHIAVMPDVH LGKGSTIGSV IPTKGAIIPA AVGVDIGCGM NALRTALTAE DLPENLAELR QAIETAVPHG RTTGRCKRDK GAWENPPVNV DAKWAELEAG YQWLTQKYPR FLNTNNYKHL GTLGTGNHFI EICLDESDQV WIMLHSGSRG IGNAIGTYFI DLAQKEMQET LETLPSRDLA YFMEGTEYFD DYLKAVAWAQ LFASLNRDAM MENVVTALQS ITQKTVRQPQ TLAMEEINCH HNYVQKEQHF GEEIYVTRKG AVSARAGQYG IIPGSMGAKS FIVRGLGNEE SFCSCSHGAG RVMSRTKAKK LFSVEDQIRA TAHVECRKDA EVIDEIPMAY KDIDAVMAAQ SDLVEVIYTL RQVVCVKG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 81% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.