missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Protein FimH Protein
A DNA sequence encoding the Escherichia coli (strain K12) Protein FimH, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7372-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_418740.1 | |
Protein FimH Protein | |
Research Use Only | |
fimH | |
Recombinant Protein | |
Yeast | |
FACKTANGTA IPIGGGSANV YVNLAPVVNV GQNLVVDLST QIFCHNDYPE TITDYVTLQR GSAYGGVLSN FSGTVKYSGS SYPFPTTSET PRVVYNSRTD KPWPVALYLT PVSSAGGVAI KAGSLIAVLI LRQTNNYNSD DFQFVWNIYA NNDVVVPTGG CDVSARDVTV TLPDYPGSVP IPLTVYCAKS QNLGYYLSGT TADAGNSIFT NTASFSPAQG VGVQLTRNGT IIPANNTVSL GAVGTSAVSL GLTANYARTG GQVTAGNVQS IIGVTFVYQ | |
Greater than 90% as determined by SDS-PAGE. |
948847 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |