missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Protein FimH Protein

A DNA sequence encoding the Escherichia coli (strain K12) Protein FimH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7372-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068530

  • 7160.00 DKK / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418740.1
Protein FimH Protein
Research Use Only
fimH
Recombinant Protein
Yeast
FACKTANGTA IPIGGGSANV YVNLAPVVNV GQNLVVDLST QIFCHNDYPE TITDYVTLQR GSAYGGVLSN FSGTVKYSGS SYPFPTTSET PRVVYNSRTD KPWPVALYLT PVSSAGGVAI KAGSLIAVLI LRQTNNYNSD DFQFVWNIYA NNDVVVPTGG CDVSARDVTV TLPDYPGSVP IPLTVYCAKS QNLGYYLSGT TADAGNSIFT NTASFSPAQG VGVQLTRNGT IIPANNTVSL GAVGTSAVSL GLTANYARTG GQVTAGNVQS IIGVTFVYQ
Greater than 90% as determined by SDS-PAGE.
948847
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers