missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant E. coli Phosphoenolpyruvate carboxylase Protein Product Code.: 16027231

enQuireBio™ Recombinant E. coli Phosphoenolpyruvate carboxylase Protein

Product Code. 16027231
50 μg, 50µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16027231

Brand: enQuireBio™ QP8934ec50ug

Quantity Price
1 3250.00 DKK / 50µg
6 3038.43 DKK / 50µg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Phosphoenolpyruvate carboxylase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_418391.1
Gene ID (Entrez) 948457
Name Phosphoenolpyruvate carboxylase Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ppc
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence YSALRSNVSM LGKVLGETIK DALGEHILER VETIRKLSKS SRAGNDANRQ ELLTTLQNLS NDELLPVARA FSQFLNLANT AEQYHSISPK GEAASNPEVI ARTLRKLKNQ PELSEDTIKK AVESLSLELV LTAHPTEITR RTLIHKMVEV NACLKQLDNK DIADYEHNQL MRRLRQLIAQ SWHTDEIRKL RPSPVDEAKW GFAVVENSLW QGVPNYLREL NEQLEENLGY KLPVEFVPVR FTSWMGGDRD GNPNVTADIT RHVLLLSRWK ATDLFLKDIQ VLVSELSMVE ATPELLALVG EEGAAEPYRY LMKNLRSRLM ATQAWLEARL KGEELPKPEG LLTQNEELWE PLYACYQSLQ ACGMGIIANG DLLDTLRRVK CFGVPLVRID IRQESTRHTE ALGELTRYLG IGDYESWSEA DKQAFLIREL NSKRPLLPRN WQPSAETREV LDTCQVIAEA PQGSIAAYVI SMAKTPSDVL AVHLLLKEAG IGFAMPVAPL FETLDDLNNA NDVMTQLLNI DWYRGLIQGK QMVMIGYSDS AKDAGVMAAS WAQYQAQDAL IKTCEKAGIE LTLFHGRGGS IGRGGAPAHA ALLSQPPGSL KGGLRVTEQG EMIRFKYGLP EITVSSLSLY TGAILEANLL PPPEPKESWR RIMDELSVIS CDVYRGYVRE NKDFVPYFRS ATPEQELGKL PLGSRPAKRR PTGGVESLRA IPWIFAWTQN RLMLPAWLGA GTALQKVVED GKQSELEAMC RDWPFFSTRL GMLEMVFAKA DLWLAEYYDQ RLVDKALWPL GKELRNLQEE DIKVVLAIAN DSHLMADLPW IAESIQLRNI YTDPLNVLQA ELLHRSRQAE KEGQEPDPRV EQALMVTIAG IAAGMRNTG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Phosphoenolpyruvate carboxylase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.