missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Mcr-1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Mcr-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7746-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_049589868.1 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Mcr-1 Protein | |
Research Use Only | |
mcr1 | |
Recombinant Protein | |
E. coli | |
MMQHTSVWYR RSVSPFVLVA SVAVFLTATA NLTFFDKISQ TYPIADNLGF VLTIAVVLFG AMLLITTLLS SYRYVLKPVL ILLLIMGAVT SYFTDTYGTV YDTTMLQNAL QTDQAETKDL LNAAFIMRII GLGVLPSLLV AFVKVDYPTW GKGLMRRLGL IVASLALILL PVVAFSSHYA SFFRVHKPLR SYVNPIMPIY SVGKLASIEY KKASAPKDTI YHAKDAVQAT KPDMRKPRLV VFVVGETARA DHVSFNGYER DTFPQLAKID GVTNFSNVTS CGTSTAYSVP CMFSYLGADE YDVDTAKYQE NVLDTLDRLG VSILWRDNNS DSKGVMDKLP KAQFADYKSA TNNAICNTNP YNECRDVGML VGLDDFVAAN NGKDMLIMLH QMGNHGPAYF KRYDEKFAKF TPVCEGNELA KCEHQSLINA YDNALLATDD FIAQSIQWLQ THSNAYDVSM LYVSDHGESL GENGVYLHGM PNAFAPKEQR SVPAFFWTDK QTGITPMATD TVLTHDAITP TLLKLFDVTA DKVKDRTAFI R | |
Greater than 90% as determined by SDS-PAGE. |