missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glycerol kinase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glycerol kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8938-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418361.1 | |
Glycerol kinase Protein | |
Research Use Only | |
glpK | |
Recombinant Protein | |
E. coli | |
TEKKYIVALD QGTTSSRAVV MDHDANIISV SQREFEQIYP KPGWVEHDPM EIWATQSSTL VEVLAKADIS SDQIAAIGIT NQRETTIVWE KETGKPIYNA IVWQCRRTAE ICEHLKRDGL EDYIRSNTGL VIDPYFSGTK VKWILDHVEG SRERARRGEL LFGTVDTWLI WKMTQGRVHV TDYTNASRTM LFNIHTLDWD DKMLEVLDIP REMLPEVRRS SEVYGQTNIG GKGGTRIPIS GIAGDQQAAL FGQLCVKEGM AKNTYGTGCF MLMNTGEKAV KSENGLLTTI ACGPTGEVNY ALEGAVFMAG ASIQWLRDEM KLINDAYDSE YFATKVQNTN GVYVVPAFTG LGAPYWDPYA RGAIFGLTRG VNANHIIRAT LESIAYQTRD VLEAMQADSG IRLHALRVDG GAVANNFLMQ FQSDILGTRV ERPEVREVTA LGAAYLAGLA VGFWQNLDEL QEKAVIEREF RPGIETTERN YRYAGWKKAV KRAMAWEEHD E | |
Greater than 90% as determined by SDS-PAGE. |
948423 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |