missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glutamate decarboxylase alpha Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glutamate decarboxylase alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6947-200ug

 View more versions of this product

Product Code. 16096820

  • 12775.00 DKK / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_417974.1
Glutamate decarboxylase alpha Protein
Research Use Only
gadA
Recombinant Protein
E. coli
MDQKLLTDFR SELLDSRFGA KAISTIAESK RFPLHEMRDD VAFQIINDEL YLDGNARQNL ATFCQTWDDE NVHKLMDLSI NKNWIDKEEY PQSAAIDLRC VNMVADLWHA PAPKNGQAVG TNTIGSSEAC MLGGMAMKWR WRKRMEAAGK PTDKPNLVCG PVQICWHKFA RYWDVELREI PMRPGQLFMD PKRMIEACDE NTIGVVPTFG VTYTGNYEFP QPLHDALDKF QADTGIDIDM HIDAASGGFL APFVAPDIVW DFRLPRVKSI SASGHKFGLA PLGCGWVIWR DEEALPQELV FNVDYLGGQI GTFAINFSRP AGQVIAQYYE FLRLGREGYT KVQNASYQVA AYLADEIAKL GPYEFICTGR PDEGIPAVCF KLKDGEDPGY TLYDLSERLR LRGWQVPAFT LGGEATDIVV MRIMCRRGFE MDFAELLLED YKASLKYLSD HPKLQGIAQQ NSFKHT
Greater than 90% as determined by SDS-PAGE.
948027
200 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Glutamate decarboxylase alpha Protein > 200μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.