missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Exopolyphosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Exopolyphosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7336-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_416997.1 | |
Exopolyphosphatase Protein | |
Research Use Only | |
ppx | |
Recombinant Protein | |
E. coli | |
PIHDKSPRPQ EFAAVDLGSN SFHMVIARVV DGAMQIIGRL KQRVHLADGL GPDNMLSEEA MTRGLNCLSL FAERLQGFSP ASVCIVGTHT LRQALNATDF LKRAEKVIPY PIEIISGNEE ARLIFMGVEH TQPEKGRKLV IDIGGGSTEL VIGENFEPIL VESRRMGCVS FAQLYFPGGV INKENFQRAR MAAAQKLETL TWQFRIQGWN VAMGASGTIK AAHEVLMEMG EKDGIITPER LEKLVKEVLR HRNFASLSLP GLSEERKTVF VPGLAILCGV FDALAIRELR LSDGALREGV LYEMEGRFRH QDVRSRTASS LANQYHIDSE QARRVLDTTM QMYEQWREQQ PKLAHPQLEA LLRWAAMLHE VGLNINHSGL HRHSAYILQN SDLPGFNQEQ QLMMATLVRY HRKAIKLDDL PRFTLFKKKQ FLPLIQLLRL GVLLNNQRQA TTTPPTLTLI TDDSHWTLRF PHDWFSQNAL VLLDLEKEQE YWEGVAGWRL KIEEESTPEI AA | |
Greater than 90% as determined by SDS-PAGE. |
946970 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Exopolyphosphatase Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction